Learn More
Meso Scale Discovery V-PLEX Mouse IL-10 Kit
SDP
Catalog No. 5000078417
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-10 Kit is an electrochemiluminescent immunoassay for measuring mouse interleukin 10 (IL-10) in various samples It uses V-PLEX technology for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for IL-10 detection antibodies with an MSD SULFO-TAG and calibrators for a standard curve The assay detects IL-10 from 0 04 pg/mL to 10 000 pg/mL
Catalog No. 50-000-78417 Supplier Meso Scale Discovery Supplier No. K152QUD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-10 Kit is an electrochemiluminescent immunoassay for measuring mouse interleukin 10 (IL-10) in various samples It uses V-PLEX technology for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for IL-10 detection antibodies with an MSD SULFO-TAG and calibrators for a standard curve The assay detects IL-10 from 0 04 pg/mL to 10 000 pg/mL