Learn More
Meso Scale Discovery V-PLEX Mouse IL-12p70 Kit
SDP
Catalog No. 5000082122
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 12 p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for a fast and sensitive chemiluminescent immunoassay with a detection range of 1 5-1000 pg/mL and a sample volume requirement of 25 L This kit can be used to measure natural and recombinant mouse IL-12p70 with high specificity and no cross-reactivity to other cytokines
Catalog No. 50-000-82122 Supplier Meso Scale Discovery Supplier No. K152QVD1
The Meso Scale Discovery V-PLEX Mouse IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 12 p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for a fast and sensitive chemiluminescent immunoassay with a detection range of 1 5-1000 pg/mL and a sample volume requirement of 25 L This kit can be used to measure natural and recombinant mouse IL-12p70 with high specificity and no cross-reactivity to other cytokines