Learn More
Meso Scale Discovery V-PLEX Mouse IL-15 Kit
SDP
Catalog No. 5000078627
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in cell culture supernatants serum plasma and tissue homogenates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 1 3 pg/mL and a detection range of 1 3-1000 pg/mL Results are obtained in 3 5 hours using the MSD QuickPlex SQ 120 instrument This assay allows for the accurate quantitation of mouse IL-15
Catalog No. 50-000-78627 Supplier Meso Scale Discovery Supplier No. K152RDD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in cell culture supernatants serum plasma and tissue homogenates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 1 3 pg/mL and a detection range of 1 3-1000 pg/mL Results are obtained in 3 5 hours using the MSD QuickPlex SQ 120 instrument This assay allows for the accurate quantitation of mouse IL-15