Learn More
Meso Scale Discovery V-PLEX Mouse IL-17A/F Kit
SDP
Catalog No. 5000082002
Encompass
The V-PLEX Mouse IL-17A/F Kit from Meso Scale Discovery is an immunoassay kit that allows for the quantitative measurement of mouse interleukin 17A and F in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plates seals The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL for IL-17A and 0 03-1250 pg/mL for IL-17F The kit can help researchers study cytokine signaling and the role of IL-17 in autoimmunity inflammation and host defense
Catalog No. 50-000-82002 Supplier Meso Scale Discovery Supplier No. K152WND1
The V-PLEX Mouse IL-17A/F Kit from Meso Scale Discovery is an immunoassay kit that allows for the quantitative measurement of mouse interleukin 17A and F in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plates seals The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL for IL-17A and 0 03-1250 pg/mL for IL-17F The kit can help researchers study cytokine signaling and the role of IL-17 in autoimmunity inflammation and host defense