Learn More
Meso Scale Discovery V-PLEX Mouse IL-1β Kit, 25 plates, 96-well, Singleplex, Sensitivity: 0.11 pg/mL, Dynamic Range: 0.11 - 1030 pg/mL, Sample Types: Serum, Plasma, Urine, Cell Culture Supernatant
SDP
Catalog No. NC1985632
Encompass
V-PLEX Mouse IL-1β Kit, 25 plates, 96-well, Singleplex, Sensitivity: 0.11 pg/mL, Dynamic Range: 0.11 - 1030 pg/mL, Sample Types: Serum, Plasma, Urine, Cell Culture Supernatant
Catalog No. NC1985632 Supplier Meso Scale Discovery Supplier No. K152QPD4
Add to Cart
Add to Cart
Request a Quote
V-PLEX Mouse IL-1β Kit, 25 plates, 96-well, Singleplex, Sensitivity: 0.11 pg/mL, Dynamic Range: 0.11 - 1030 pg/mL, Sample Types: Serum, Plasma, Urine, Cell Culture Supernatant