Learn More
Meso Scale Discovery V-PLEX Mouse IL-2 Kit
SDP
Catalog No. 5000078355
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a plate coated with a mouse IL-2 specific capture antibody a ruthenium labeled mouse IL-2 specific detection antibody and calibrators and controls to generate a standard curve and validate assay performance The assay has a detection range from 3 2 to 10 000 pg/mL and exhibits no cross-reactivity with other cytokines The kit includes reagents for one 96-well plate and can test up to 40 samples in duplicate
Catalog No. 50-000-78355 Supplier Meso Scale Discovery Supplier No. K152QQD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a plate coated with a mouse IL-2 specific capture antibody a ruthenium labeled mouse IL-2 specific detection antibody and calibrators and controls to generate a standard curve and validate assay performance The assay has a detection range from 3 2 to 10 000 pg/mL and exhibits no cross-reactivity with other cytokines The kit includes reagents for one 96-well plate and can test up to 40 samples in duplicate