Learn More
Meso Scale Discovery V-PLEX Mouse IL-2 Kit
SDP
Catalog No. 5000080440
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with capture antibodies against mouse IL-2 detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant mouse IL-2 The assay has a detection range of 3 2-10 000 pg/mL and requires 2 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of mouse IL-2 levels
Catalog No. 50-000-80440 Supplier Meso Scale Discovery Supplier No. K152QQD1
The Meso Scale Discovery V-PLEX Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with capture antibodies against mouse IL-2 detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant mouse IL-2 The assay has a detection range of 3 2-10 000 pg/mL and requires 2 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of mouse IL-2 levels