Learn More
Meso Scale Discovery V-PLEX Mouse IL-33 Kit
SDP
Catalog No. 5000079061
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-33 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It uses MSD s V-PLEX technology offering high sensitivity a broad dynamic range up to 3 logs and no cross-reactivity with other cytokines The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and a 10X Read Buffer for chemiluminescent signal measurement by MSD s SECTOR Imager It quantifies IL-33 from 1 5 to 1000 pg/mL with a 50 L sample
Catalog No. 50-000-79061 Supplier Meso Scale Discovery Supplier No. K152XBD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-33 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It uses MSD s V-PLEX technology offering high sensitivity a broad dynamic range up to 3 logs and no cross-reactivity with other cytokines The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and a 10X Read Buffer for chemiluminescent signal measurement by MSD s SECTOR Imager It quantifies IL-33 from 1 5 to 1000 pg/mL with a 50 L sample