Learn More
Meso Scale Discovery V-PLEX Mouse IL-33 Kit
SDP
Catalog No. 5000080319
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-33 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay takes 3 5 hours to complete and each kit contains sufficient reagents for 1 x 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-80319 Supplier Meso Scale Discovery Supplier No. K152XBD1
The Meso Scale Discovery V-PLEX Mouse IL-33 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay takes 3 5 hours to complete and each kit contains sufficient reagents for 1 x 96-well plate and can assay 40 samples in duplicate