Learn More
Meso Scale Discovery V-PLEX Mouse IL-4 Kit
SDP
Catalog No. 5000079315
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 4 (IL-4) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-10 000 pg/mL It uses an ECL-based method for high sensitivity and low background taking about 3 5 hours to complete with enough reagents for one 96-well plate
Catalog No. 50-000-79315 Supplier Meso Scale Discovery Supplier No. K152QRD1
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 4 (IL-4) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-10 000 pg/mL It uses an ECL-based method for high sensitivity and low background taking about 3 5 hours to complete with enough reagents for one 96-well plate