Learn More
Meso Scale Discovery V-PLEX Mouse IL-9 Kit
SDP
Catalog No. 5000080340
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 3 2-10 000 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit includes sufficient reagents for 1 x 96-well plate when run as described in the product insert
Catalog No. 50-000-80340 Supplier Meso Scale Discovery Supplier No. K152XCD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 3 2-10 000 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit includes sufficient reagents for 1 x 96-well plate when run as described in the product insert