Learn More
Meso Scale Discovery V-PLEX Mouse IL-9 Kit
SDP
Catalog No. 5000078692
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 3 2-10 000 pg/mL and includes reagents for testing 50 microplate wells This multiplex assay allows for the simultaneous measurement of mouse IL-9 with other biomarkers in a single sample
Catalog No. 50-000-78692 Supplier Meso Scale Discovery Supplier No. K152XCD1
The Meso Scale Discovery V-PLEX Mouse IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 3 2-10 000 pg/mL and includes reagents for testing 50 microplate wells This multiplex assay allows for the simultaneous measurement of mouse IL-9 with other biomarkers in a single sample