Learn More
Meso Scale Discovery V-PLEX Mouse IL-9 Kit
SDP
Supplier:  Meso Scale Discovery K152XCD2
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 9 (IL-9) in cell culture supernatants serum plasma and tissue lysates The assay utilizes MSD s proprietary SULFO-TAG labels which emit light when electrochemically stimulated via carbon electrodes in the bottom of each microwell The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-82261