Learn More
Meso Scale Discovery V-PLEX Mouse IP-10 Kit
SDP
Catalog No. 5000081454
Encompass
The Meso Scale Discovery V-PLEX Mouse IP-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interferon gamma-induced protein 10 (IP-10) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IP-10 with a lower limit of detection of 1 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with IP-10 capture antibody detection antibody calibrator diluent wash buffer read buffer and a plate seal The assay can be fully automated on MSD instrument platforms and has a total assay time of 3 5 hours
Catalog No. 50-000-81454 Supplier Meso Scale Discovery Supplier No. K152NVD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IP-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interferon gamma-induced protein 10 (IP-10) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IP-10 with a lower limit of detection of 1 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with IP-10 capture antibody detection antibody calibrator diluent wash buffer read buffer and a plate seal The assay can be fully automated on MSD instrument platforms and has a total assay time of 3 5 hours