Learn More
Meso Scale Discovery V-PLEX Mouse KC/GRO Kit
SDP
Catalog No. 5000078703
Encompass
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent wash buffer and standards to generate a standard curve The electrochemiluminescent detection method provides high sensitivity wide dynamic range and requires low sample volume This product can aid in understanding immune response and inflammation in mouse models
Catalog No. 50-000-78703 Supplier Meso Scale Discovery Supplier No. K152QTD1
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent wash buffer and standards to generate a standard curve The electrochemiluminescent detection method provides high sensitivity wide dynamic range and requires low sample volume This product can aid in understanding immune response and inflammation in mouse models