Learn More
Meso Scale Discovery V-PLEX Mouse KC/GRO Kit
SDP
Catalog No. 5000078704
Encompass
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized KC and GRO standards to generate standard curves for quantification The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can be used to study inflammatory responses in mouse models of human disease
Catalog No. 50-000-78704 Supplier Meso Scale Discovery Supplier No. K152QTD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized KC and GRO standards to generate standard curves for quantification The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can be used to study inflammatory responses in mouse models of human disease