Learn More
Meso Scale Discovery V-PLEX Mouse KC/GRO Kit
SDP
Catalog No. 5000079099
Encompass
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Calibrator Diluent wash buffer and standards and controls The assay takes about 3 5 hours and detects KC at 1 53-2500 pg/mL and GRO at 0 61-1000 pg/mL using MSD s SECTOR Imager for high sensitivity and a wide dynamic range
Catalog No. 50-000-79099 Supplier Meso Scale Discovery Supplier No. K152QTD2
The V-PLEX Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Calibrator Diluent wash buffer and standards and controls The assay takes about 3 5 hours and detects KC at 1 53-2500 pg/mL and GRO at 0 61-1000 pg/mL using MSD s SECTOR Imager for high sensitivity and a wide dynamic range