Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery V-PLEX Mouse MCP-1 Kit, 96-well, Electrochemiluminescence (ECL), Mouse MCP-1 (CCL2) detection, LLOD 0.672 pg/mL, Dynamic range 0.672 - 325 pg/mL, For Research Use Only
SDP

Catalog No. NC2887947
Encompass

The MESO SCALE DISCOVERY V-PLEX Mouse MCP-1 Kit is an advanced immunoassay designed for quantitative measurement of Mouse MCP-1 (CCL2) levels, ensuring consistent and reproducible results across different testing conditions. This kit is ideal for researchers focusing on immunology and cytokine studies, requiring robust tools for effective detection of MCP-1 in various biological samples.

  • Analyte: MCP-1 (CCL2)
  • Species: Mouse
  • Plate type: 96-well
  • Detection method: Electrochemiluminescence (ECL)
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM, SECTOR Imager 6000
  • Capture antibody: Rat Monoclonal
  • Detection antibody: Goat Polyclonal
  • Lower limit of detection (LLOD): 0.672 pg/mL
  • Dynamic range: 0.672 - 325 pg/mL
  • Sample types: Serum, Plasma, Cell Culture Supernatant, Urine
  • Storage condition: Multi-Component

Catalog No. NC2887947 Supplier Meso Scale Discovery Supplier No. K152NND2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY V-PLEX Mouse MCP-1 Kit is an advanced immunoassay designed for quantitative measurement of Mouse MCP-1 (CCL2) levels, ensuring consistent and reproducible results across different testing conditions. This kit is ideal for researchers focusing on immunology and cytokine studies, requiring robust tools for effective detection of MCP-1 in various biological samples.

  • Analyte: MCP-1 (CCL2)
  • Species: Mouse
  • Plate type: 96-well
  • Detection method: Electrochemiluminescence (ECL)
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM, SECTOR Imager 6000
  • Capture antibody: Rat Monoclonal
  • Detection antibody: Goat Polyclonal
  • Lower limit of detection (LLOD): 0.672 pg/mL
  • Dynamic range: 0.672 - 325 pg/mL
  • Sample types: Serum, Plasma, Cell Culture Supernatant, Urine
  • Storage condition: Multi-Component

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.