Learn More
Meso Scale Discovery V-PLEX Mouse MCP-1 Kit
SDP
Catalog No. 5000081494
Encompass
The V-PLEX Mouse MCP-1 Kit from Meso Scale Discovery measures mouse MCP-1 in serum plasma cell culture supernatants and lavage fluids This electrochemiluminescent immunoassay uses SULFO-TAG labels and multi-array technology for sensitive detection with a range of 1 3 to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments enabling quantitative measurement of mouse MCP-1 in research
Catalog No. 50-000-81494 Supplier Meso Scale Discovery Supplier No. K152NND1
Add to Cart
Add to Cart
The V-PLEX Mouse MCP-1 Kit from Meso Scale Discovery measures mouse MCP-1 in serum plasma cell culture supernatants and lavage fluids This electrochemiluminescent immunoassay uses SULFO-TAG labels and multi-array technology for sensitive detection with a range of 1 3 to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments enabling quantitative measurement of mouse MCP-1 in research