Learn More
Meso Scale Discovery V-PLEX Mouse MIP-1 Kit
SDP
Catalog No. 5000081511
Encompass
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1 alpha in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and takes approximately 3 5 hours to complete It utilizes electrochemiluminescence for high sensitivity and wide dynamic range This product can aid in understanding immune responses and inflammation
Catalog No. 50-000-81511 Supplier Meso Scale Discovery Supplier No. K152NQD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1 alpha in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and takes approximately 3 5 hours to complete It utilizes electrochemiluminescence for high sensitivity and wide dynamic range This product can aid in understanding immune responses and inflammation