Learn More
Meso Scale Discovery V-PLEX Mouse MIP-1 Kit
SDP
Catalog No. 5000082108
Encompass
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is used to quantitatively measure mouse MIP-1 alpha (CCL3) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of mouse MIP-1 alpha on the Meso QuickPlex SQ 120 instrument The kit includes a mouse MIP-1 alpha calibrator and pre-coated plates to enable accurate quantitation of mouse MIP-1 alpha concentrations within 90 minutes
Catalog No. 50-000-82108 Supplier Meso Scale Discovery Supplier No. K152NQD1
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is used to quantitatively measure mouse MIP-1 alpha (CCL3) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of mouse MIP-1 alpha on the Meso QuickPlex SQ 120 instrument The kit includes a mouse MIP-1 alpha calibrator and pre-coated plates to enable accurate quantitation of mouse MIP-1 alpha concentrations within 90 minutes