Learn More
Meso Scale Discovery V-PLEX Mouse MIP-2 Kit
SDP
Catalog No. 5000080041
Encompass
The V-PLEX Mouse MIP-2 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse MIP-2 in various samples using electrochemiluminescence It includes a 96-well plate with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and lyophilized calibrator for a standard curve The assay detects 3 2-10 000 pg/mL and tests up to 40 samples in duplicate
Catalog No. 50-000-80041 Supplier Meso Scale Discovery Supplier No. K152QCD2
The V-PLEX Mouse MIP-2 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse MIP-2 in various samples using electrochemiluminescence It includes a 96-well plate with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and lyophilized calibrator for a standard curve The assay detects 3 2-10 000 pg/mL and tests up to 40 samples in duplicate