Learn More
Meso Scale Discovery V-PLEX Mouse MIP-2 Kit
SDP
Catalog No. 5000081456
Encompass
The Meso Scale Discovery V-PLEX Mouse MIP-2 Kit is used to quantitatively measure mouse macrophage inflammatory protein 2 (MIP-2) in various biological samples such as serum plasma cell culture supernatants and lavage fluids This electrochemiluminescence (ECL) based immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and reproducible results comparable to ELISA The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The kit can accurately detect mouse MIP-2 concentrations between 1 5-1000 pg/mL with an assay time of 3 5 hours
Catalog No. 50-000-81456 Supplier Meso Scale Discovery Supplier No. K152QCD1
The Meso Scale Discovery V-PLEX Mouse MIP-2 Kit is used to quantitatively measure mouse macrophage inflammatory protein 2 (MIP-2) in various biological samples such as serum plasma cell culture supernatants and lavage fluids This electrochemiluminescence (ECL) based immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and reproducible results comparable to ELISA The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The kit can accurately detect mouse MIP-2 concentrations between 1 5-1000 pg/mL with an assay time of 3 5 hours