Learn More
Meso Scale Discovery V-PLEX Mouse TNF-alpha Kit, 5 plates
SDP
Catalog No. NC1177030
Encompass
The Mouse TNF-alpha Kit provides assay-specific components for the quantitative determination of endogenous TNF-alpha in mouse urine, serum, plasma, and cell culture supernatants.
Catalog No. NC1177030 Supplier Meso Scale Discovery Supplier No. K152QWD2
Add to Cart
Add to Cart
The Mouse TNF-alpha Kit provides assay-specific components for the quantitative determination of endogenous TNF-alpha in mouse urine, serum, plasma, and cell culture supernatants.