Learn More
Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit
SDP
Catalog No. 5000078554
Encompass
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures Eotaxin-3 levels in non-human primate serum plasma and culture supernatants The kit uses electrochemiluminescence detection technology and provides accurate reproducible results in just 3 5 hours The V-PLEX platform has high sensitivity a broad dynamic range and requires just 12 5 L or 25 L sample This Eotaxin-3 assay kit can be used to monitor immune responses and inflammation in non-human primate studies
Catalog No. 50-000-78554 Supplier Meso Scale Discovery Supplier No. K156NUD2
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures Eotaxin-3 levels in non-human primate serum plasma and culture supernatants The kit uses electrochemiluminescence detection technology and provides accurate reproducible results in just 3 5 hours The V-PLEX platform has high sensitivity a broad dynamic range and requires just 12 5 L or 25 L sample This Eotaxin-3 assay kit can be used to monitor immune responses and inflammation in non-human primate studies