Learn More
Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit
SDP
Catalog No. 5000081278
Encompass
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin-3 in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This product can aid in the research of inflammatory and immunological conditions related to Eotaxin-3
Catalog No. 50-000-81278 Supplier Meso Scale Discovery Supplier No. K156NUD1
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin-3 in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This product can aid in the research of inflammatory and immunological conditions related to Eotaxin-3