Learn More
Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit
SDP
Catalog No. 5000081385
Encompass
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures Eotaxin-3 levels in non-human primate serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires just 50 L sample volume per well The V-PLEX Eotaxin-3 Kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluent The kit can accurately quantify Eotaxin-3 concentrations from 0 04 - 500 pg/mL with an inter-assay CV of 12%
Catalog No. 50-000-81385 Supplier Meso Scale Discovery Supplier No. K156NUD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures Eotaxin-3 levels in non-human primate serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires just 50 L sample volume per well The V-PLEX Eotaxin-3 Kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluent The kit can accurately quantify Eotaxin-3 concentrations from 0 04 - 500 pg/mL with an inter-assay CV of 12%