Learn More
Meso Scale Discovery V-PLEX NHP GM-CSF Kit
SDP
Catalog No. 5000081391
Encompass
The V-PLEX NHP GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human GM-CSF in CSF It uses ECL detection for high sensitivity broad range and low sample volume The assay measures GM-CSF and other biomarkers simultaneously The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and seals It aids in understanding GM-CSF biology and its role in CNS diseases
Catalog No. 50-000-81391 Supplier Meso Scale Discovery Supplier No. K156RID2
The V-PLEX NHP GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human GM-CSF in CSF It uses ECL detection for high sensitivity broad range and low sample volume The assay measures GM-CSF and other biomarkers simultaneously The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and seals It aids in understanding GM-CSF biology and its role in CNS diseases