Learn More
Meso Scale Discovery V-PLEX NHP IFN-y Kit
SDP
Catalog No. 5000078556
Encompass
The V-PLEX NHP IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for measuring natural human interferon gamma (IFN-gamma) in various biological fluids It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrators for a standard curve The assay detects IFN-gamma from 0 05 pg/mL to 10 000 pg/mL
Catalog No. 50-000-78556 Supplier Meso Scale Discovery Supplier No. K156QOD1
The V-PLEX NHP IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for measuring natural human interferon gamma (IFN-gamma) in various biological fluids It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrators for a standard curve The assay detects IFN-gamma from 0 05 pg/mL to 10 000 pg/mL