Learn More
Meso Scale Discovery V-PLEX NHP IFN-y Kit
SDP
Catalog No. 5000081386
Encompass
The V-PLEX NHP IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of natural and recombinant non-human primate interferon gamma in serum plasma cell culture supernatants and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescence immunoassay has high sensitivity a broad dynamic range and requires only 25 L of sample The kit can help researchers study immune system function and response in non-human primate models
Catalog No. 50-000-81386 Supplier Meso Scale Discovery Supplier No. K156QOD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of natural and recombinant non-human primate interferon gamma in serum plasma cell culture supernatants and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescence immunoassay has high sensitivity a broad dynamic range and requires only 25 L of sample The kit can help researchers study immune system function and response in non-human primate models