Learn More
Meso Scale Discovery V-PLEX NHP IFN-γ Kit, 5 plates, 96-well plate format, Single-analyte, Serum, Plasma, Urine, Cell Culture Supernatant, LLOD 0.37 pg/mL, Dynamic Range 0.37 - 938 pg/mL
SDP
Catalog No. NC3593591
Encompass
V-PLEX NHP IFN-γ Kit, 5 plates, 96-well plate format, Single-analyte, Serum, Plasma, Urine, Cell Culture Supernatant, LLOD 0.37 pg/mL, Dynamic Range 0.37 - 938 pg/mL
Catalog No. NC3593591 Supplier Meso Scale Discovery Supplier No. K156QOD2
V-PLEX NHP IFN-γ Kit, 5 plates, 96-well plate format, Single-analyte, Serum, Plasma, Urine, Cell Culture Supernatant, LLOD 0.37 pg/mL, Dynamic Range 0.37 - 938 pg/mL