Learn More
Meso Scale Discovery V-PLEX NHP IL-10 Kit
SDP
Catalog No. 5000079021
Encompass
The Meso Scale Discovery V-PLEX NHP IL-10 Kit is a sensitive immunoassay for measuring non-human primate IL-10 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity and a wide dynamic range requiring just 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an optimized assay diluent It quantifies NHP IL-10 from 15 6 to 1000 pg/mL with an inter-assay CV of 10%
Catalog No. 50-000-79021 Supplier Meso Scale Discovery Supplier No. K156QUD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-10 Kit is a sensitive immunoassay for measuring non-human primate IL-10 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity and a wide dynamic range requiring just 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an optimized assay diluent It quantifies NHP IL-10 from 15 6 to 1000 pg/mL with an inter-assay CV of 10%