Learn More
Meso Scale Discovery V-PLEX NHP IL-10 Kit
SDP
Catalog No. 5000079943
Encompass
The Meso Scale Discovery V-PLEX NHP IL-10 Kit is a highly sensitive immunoassay that enables the quantitative measurement of non-human primate interleukin 10 (IL-10) in serum plasma and cell culture supernatants The electrochemiluminescent detection technology provides high sensitivity broad dynamic range and requires low sample volume This kit includes a plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79943 Supplier Meso Scale Discovery Supplier No. K156QUD1
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX NHP IL-10 Kit is a highly sensitive immunoassay that enables the quantitative measurement of non-human primate interleukin 10 (IL-10) in serum plasma and cell culture supernatants The electrochemiluminescent detection technology provides high sensitivity broad dynamic range and requires low sample volume This kit includes a plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL