Learn More
Meso Scale Discovery V-PLEX NHP IL-10 Kit
SDP
Catalog No. 5000081525
Encompass
The V-PLEX NHP IL-10 Kit from Meso Scale Discovery is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-10 in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with a monoclonal antibody specific for human IL-10 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a detection range of 0 06-500 pg/mL and exhibits minimal cross-reactivity with other cytokines This kit allows for rapid and accurate quantification of natural human IL-10 levels in biological samples
Catalog No. 50-000-81525 Supplier Meso Scale Discovery Supplier No. K156QUD2
The V-PLEX NHP IL-10 Kit from Meso Scale Discovery is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-10 in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with a monoclonal antibody specific for human IL-10 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a detection range of 0 06-500 pg/mL and exhibits minimal cross-reactivity with other cytokines This kit allows for rapid and accurate quantification of natural human IL-10 levels in biological samples