Learn More
Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit
SDP
Catalog No. 5000081112
Encompass
The Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-12 and IL-23 in serum plasma and culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are analyzed on MSD s SECTOR Imager for high sensitivity and a broad dynamic range The kit allows for the precise quantification of IL-12 and IL-23 cytokines
Catalog No. 50-000-81112 Supplier Meso Scale Discovery Supplier No. K156RJD1
The Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-12 and IL-23 in serum plasma and culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are analyzed on MSD s SECTOR Imager for high sensitivity and a broad dynamic range The kit allows for the precise quantification of IL-12 and IL-23 cytokines