Learn More
Meso Scale Discovery V-PLEX NHP IL-15 Kit 25Plate
SDP
Catalog No. 5000079937
Encompass
The Meso Scale Discovery V-PLEX NHP IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 2 pg/mL and a range from 0 53 pg/mL to 1000 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate
Catalog No. 50-000-79937 Supplier Meso Scale Discovery Supplier No. K156RDD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 2 pg/mL and a range from 0 53 pg/mL to 1000 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate