Learn More
Meso Scale Discovery V-PLEX NHP IL-15 Kit
SDP
Catalog No. 5000080808
Encompass
The V-PLEX NHP IL-15 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It includes a 96-well plate with an IL-15-specific monoclonal antibody detection antibody solution buffers diluent and calibrator The assay s sensitivity is 0 3 pg/mL with a calibration curve from 3 2 to 10 000 pg/mL The electrochemiluminescent method offers high sensitivity wide range and low sample volume requirements enabling reliable IL-15 quantification in human samples
Catalog No. 50-000-80808 Supplier Meso Scale Discovery Supplier No. K156RDD1
The V-PLEX NHP IL-15 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It includes a 96-well plate with an IL-15-specific monoclonal antibody detection antibody solution buffers diluent and calibrator The assay s sensitivity is 0 3 pg/mL with a calibration curve from 3 2 to 10 000 pg/mL The electrochemiluminescent method offers high sensitivity wide range and low sample volume requirements enabling reliable IL-15 quantification in human samples