Learn More
Meso Scale Discovery V-PLEX NHP IL-15 Kit
SDP
Catalog No. 5000081388
Encompass
The V-PLEX NHP IL-15 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-15 in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with a capture antibody specific for human IL-15 a detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay The total assay time is 3 5 hours This kit allows for the accurate quantification of IL-15 levels in human biological samples
Catalog No. 50-000-81388 Supplier Meso Scale Discovery Supplier No. K156RDD2
The V-PLEX NHP IL-15 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-15 in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with a capture antibody specific for human IL-15 a detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay The total assay time is 3 5 hours This kit allows for the accurate quantification of IL-15 levels in human biological samples