Learn More
Meso Scale Discovery V-PLEX NHP IL-16 Kit
SDP
Catalog No. 5000079932
Encompass
The Meso Scale Discovery V-PLEX NHP IL-16 Kit is a sensitive immunoassay for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 instrument offering a reproducible method to measure IL-16 levels in biological samples
Catalog No. 50-000-79932 Supplier Meso Scale Discovery Supplier No. K156RED2
The Meso Scale Discovery V-PLEX NHP IL-16 Kit is a sensitive immunoassay for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 instrument offering a reproducible method to measure IL-16 levels in biological samples