Learn More
Meso Scale Discovery V-PLEX NHP IL-17A Kit
SDP
Catalog No. 5000079964
Encompass
The V-PLEX NHP IL-17A Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-17A in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 4 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with IL-17A capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay can be read on MSD s SECTOR Imager for results in as little as 3 5 hours
Catalog No. 50-000-79964 Supplier Meso Scale Discovery Supplier No. K156RFD1
The V-PLEX NHP IL-17A Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-17A in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 4 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with IL-17A capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay can be read on MSD s SECTOR Imager for results in as little as 3 5 hours