Learn More
Meso Scale Discovery V-PLEX NHP IL-17A Kit
SDP
Catalog No. 5000081111
Encompass
The V-PLEX NHP IL-17A Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-17A in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody to human IL-17A a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-81111 Supplier Meso Scale Discovery Supplier No. K156RFD2
The V-PLEX NHP IL-17A Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-17A in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody to human IL-17A a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL Sample incubation time is 2 hours