Learn More
Meso Scale Discovery V-PLEX NHP IL-1SY Kit
SDP
Catalog No. 5000079998
Encompass
The Meso Scale Discovery V-PLEX NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-400 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit provides a simple rapid and sensitive method to quantify IL-1 levels
Catalog No. 50-000-79998 Supplier Meso Scale Discovery Supplier No. K156QPD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-400 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit provides a simple rapid and sensitive method to quantify IL-1 levels