Learn More
Meso Scale Discovery V-PLEX NHP IL-2 Kit
SDP
Catalog No. 5000078553
Encompass
The Meso Scale Discovery V-PLEX NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 2 (IL-2) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL Sample incubation time is 2 hours The plate is read on the MSD instrument for quantitative analysis This IL-2 kit provides sensitive precise and reproducible quantitation of IL-2 levels
Catalog No. 50-000-78553 Supplier Meso Scale Discovery Supplier No. K156QQD1
The Meso Scale Discovery V-PLEX NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 2 (IL-2) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL Sample incubation time is 2 hours The plate is read on the MSD instrument for quantitative analysis This IL-2 kit provides sensitive precise and reproducible quantitation of IL-2 levels