Learn More
Meso Scale Discovery V-PLEX NHP IL-2 Kit
SDP
Catalog No. 5000081387
Encompass
The V-PLEX NHP IL-2 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-2 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a 7-point calibration curve from 0 06 to 600 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate when used as directed
Catalog No. 50-000-81387 Supplier Meso Scale Discovery Supplier No. K156QQD2
The V-PLEX NHP IL-2 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-2 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a 7-point calibration curve from 0 06 to 600 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate when used as directed