Learn More
Meso Scale Discovery V-PLEX NHP IL-5 Kit
SDP
Catalog No. 5000079948
Encompass
The V-PLEX NHP IL-5 Kit from Meso Scale Discovery is an immunoassay that quantitatively measures natural and recombinant human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-5 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and a 7-log dynamic range from 0 08-600 pg/mL It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate
Catalog No. 50-000-79948 Supplier Meso Scale Discovery Supplier No. K156QSD1
The V-PLEX NHP IL-5 Kit from Meso Scale Discovery is an immunoassay that quantitatively measures natural and recombinant human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-5 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and a 7-log dynamic range from 0 08-600 pg/mL It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate