Learn More
Meso Scale Discovery V-PLEX NHP IL-5 Kit 25Plate
SDP
Catalog No. 5000081277
Encompass
The Meso Scale Discovery V-PLEX NHP IL-5 Kit is a highly sensitive immunoassay that enables the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires low sample volumes The V-PLEX kit includes a 96-well plate coated with an IL-5 capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators and a diluent The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81277 Supplier Meso Scale Discovery Supplier No. K156QSD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-5 Kit is a highly sensitive immunoassay that enables the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires low sample volumes The V-PLEX kit includes a 96-well plate coated with an IL-5 capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators and a diluent The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL