Learn More
Meso Scale Discovery V-PLEX NHP IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K156QXD4
Encompass
The Meso Scale Discovery V-PLEX NHP IL-6 Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 6 (IL-6) in serum plasma and culture supernatants It detects 0 06-1500 pg/mL using 50 L or less sample volume The kit includes a 96-well plate with IL-6 capture antibody detection antibody with MSD SULFO-TAG calibrators and necessary reagents It offers high sensitivity wide dynamic range and no wash steps Results are available in 3 5 hours with an MSD instrument This assay provides rapid ultra-sensitive and robust quantitative measurement
Catalog No. 50-000-79022
Add to Cart
Request a Quote