Learn More
Meso Scale Discovery V-PLEX NHP IL-6 Kit
SDP
Catalog No. 5000080804
Encompass
The Meso Scale Discovery V-PLEX NHP IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-80804 Supplier Meso Scale Discovery Supplier No. K156QXD1
The Meso Scale Discovery V-PLEX NHP IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a capture antibody specific for IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate