Learn More
Meso Scale Discovery V-PLEX NHP IL-7 Kit
SDP
Catalog No. 5000080806
Encompass
The Meso Scale Discovery V-PLEX NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit contains a plate coated with a monoclonal antibody to human IL-7 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-7 levels
Catalog No. 50-000-80806 Supplier Meso Scale Discovery Supplier No. K156RCD1
The Meso Scale Discovery V-PLEX NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit contains a plate coated with a monoclonal antibody to human IL-7 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-7 levels