Learn More
Meso Scale Discovery V-PLEX NHP IL-7 Kit
SDP
Catalog No. 5000080807
Encompass
The Meso Scale Discovery V-PLEX NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with IL-7 capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The lower limit of detection is 0 10 pg/mL with an upper limit of 1000 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80807 Supplier Meso Scale Discovery Supplier No. K156RCD2
The Meso Scale Discovery V-PLEX NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with IL-7 capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The lower limit of detection is 0 10 pg/mL with an upper limit of 1000 pg/mL The assay takes approximately 3 5 hours to complete